Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, dNTPs

VWR®, dNTPs

Set dNTP (dATP, dCTP, dGTP, dTTP) 100mM of each (8 × 10ml), 80 mL
Catalog # 733-2846
Supplier:   VWR Chemicals
VWR®, dNTPs
Catalog # 733-2846
Supplier:  VWR Chemicals
CAS Number:  
VWR®, dNTPs
VWR®, dNTPs

Set dNTP (dATP, dCTP, dGTP, dTTP) 100mM of each (8 × 10ml), 80 mL

CAS Number:  
Price
7 580,00 €
Log in for availability
1 * 80 mL
Product Details & Documents

Ready to use molecular biology grade dNTP mixes and dNTP sets. The dNTP mix is designed to save hands-on time for researchers, and reduce the possibility of contamination by reducing pipetting. dNTP solutions are also available in sets of four individual dNTPs, each 100 mm. Both are convenient for use in DNA polymerisation reactions, DNA labelling and sequencing processes.

  • Available as pre-mixed 10 or 25 mM solutions, or as sets of individual 100 mM dNTP solutions
  • Both pre-mixed and sets have been functionally tested in PCR
  • Purity >99% by HPLC
  • Supplied in solution at pH 7,3 to 7,5
Specifications

Specifications

  • Pk:
    80 mL
  • Description:
    Set dNTP (dATP, dCTP, dGTP, dTTP) 100mM of each (8 × 10ml)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.