ContactUsLinkComponent
Minimum Essential Media Description: -Indicator: Phenol Red -Supplementation: standard serum -Supplementation HEPES buffer: NO HEPPES buffer -Glocose: Low -pack size: 500ml 1 * 500 mL
Catalog # HIMEAL047A-500ML
Supplier:  HIMEDIA
Minimum Essential Media Description: -Indicator: Phenol Red -Supplementation: standard serum -Supplementation HEPES buffer: NO HEPPES buffer -Glocose: Low -pack size: 500ml 1 * 500 mL
Catalog # HIMEAL047A-500ML
Supplier:  HIMEDIA
Frequently Bought Together