ContactUsLinkComponent
Counting chambers (Thoma ruling with special depth), Helber, depth 0.02 mm, double net ruling, dark line, without clamps, with two cover glasses of 0.4 mm thickness, MARIENFELD SUPERIOR 1 * 1 items
Catalog # MARI0642710
Supplier:  MARIENFELD
Counting chambers (Thoma ruling with special depth), Helber, depth 0.02 mm, double net ruling, dark line, without clamps, with two cover glasses of 0.4 mm thickness, MARIENFELD SUPERIOR 1 * 1 items
Catalog # MARI0642710
Supplier:  MARIENFELD
Frequently Bought Together