ContactUsLinkComponent
Cynomolgus Rhesus macaque CD47, Fc Tag (CD7-C5252) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 141 (Acce ssion F7A802-1). In the region Gln 19 - Glu 141, the AA sequence of Cynomolgus and Rhesus macaque CD47 are homologus. 1 * 1 mg
Catalog # ACRBCD7-C5252-1MG
Supplier:  ACROBIOSYSTEMS
Cynomolgus Rhesus macaque CD47, Fc Tag (CD7-C5252) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 141 (Acce ssion F7A802-1). In the region Gln 19 - Glu 141, the AA sequence of Cynomolgus and Rhesus macaque CD47 are homologus. 1 * 1 mg
Catalog # ACRBCD7-C5252-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together