ContactUsLinkComponent
Recombinat Protein,Biotinylated Human VSIG8 Protein, Fc,Avitag™,Biotinylated Human VSIG8, Fc,Avitag (VS8-H82F2) is expressed from hu man 293 cells (HEK293). It contains AA Val 22 - Gly 263 (Accession NP001013683.1).This protein carries a human IgG1 F 1 * 25 µG
Catalog # ACRBVS8-H82F2-25UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human VSIG8 Protein, Fc,Avitag™,Biotinylated Human VSIG8, Fc,Avitag (VS8-H82F2) is expressed from hu man 293 cells (HEK293). It contains AA Val 22 - Gly 263 (Accession NP001013683.1).This protein carries a human IgG1 F 1 * 25 µG
Catalog # ACRBVS8-H82F2-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together