ContactUsLinkComponent
Recombinat Protein,Human Bcl-x BCL2L1 Protein, His Tag,Human Bcl-x, His Tag (BC1-H5120) is expressed from E.coli cells. It contains AA Met 1 - Arg 212 (Accession AAH19307).This protein carries a polyhistidine tag at the C-terminus. The protein has a 1 * 1 mg
Catalog # ACRBBC1-H5120-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Bcl-x BCL2L1 Protein, His Tag,Human Bcl-x, His Tag (BC1-H5120) is expressed from E.coli cells. It contains AA Met 1 - Arg 212 (Accession AAH19307).This protein carries a polyhistidine tag at the C-terminus. The protein has a 1 * 1 mg
Catalog # ACRBBC1-H5120-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together