ContactUsLinkComponent
Biotinylated Human CD38, Avitag,His Tag (CD8-H82E7) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Acc ession NP001766.2). 1 * 200 µG
Catalog # ACRBCD8-H82E7200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human CD38, Avitag,His Tag (CD8-H82E7) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Acc ession NP001766.2). 1 * 200 µG
Catalog # ACRBCD8-H82E7200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together