ContactUsLinkComponent
Biotinylated Human ROR1, Fc,Avitag (RO1-H82F4) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accessio n Q01973-1). 1 * 200 µG
Catalog # ACRBRO1-H82F4200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human ROR1, Fc,Avitag (RO1-H82F4) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accessio n Q01973-1). 1 * 200 µG
Catalog # ACRBRO1-H82F4200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together