ContactUsLinkComponent
Human TGFBR2, Fc Tag (TG2-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Asp 159 (Accession NP003233). 1 * 1 mg
Catalog # ACRBTG2-H5252-1MG
Supplier:  ACROBIOSYSTEMS
Human TGFBR2, Fc Tag (TG2-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Asp 159 (Accession NP003233). 1 * 1 mg
Catalog # ACRBTG2-H5252-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together