ContactUsLinkComponent
Biotinylated Mouse OX40, His,Avitag (OX0-M82E5) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 211 (Accessi on P47741-1). 1 * 200 µG
Catalog # ACRBOX0-M82E5200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Mouse OX40, His,Avitag (OX0-M82E5) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 211 (Accessi on P47741-1). 1 * 200 µG
Catalog # ACRBOX0-M82E5200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together