ContactUsLinkComponent
Cynomolgus Rhesus macaque BCMA, His Tag (BCA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Acces sion G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus. 1 * 1 mg
Catalog # ACRBBCA-C52H7-1MG
Supplier:  ACROBIOSYSTEMS
Cynomolgus Rhesus macaque BCMA, His Tag (BCA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Acces sion G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus. 1 * 1 mg
Catalog # ACRBBCA-C52H7-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together