ContactUsLinkComponent
Cynomolgus Her2, His Tag (HE2-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession XP005584 091.2). 1 * 100 µG
Catalog # ACRBHE2-C52HB100UG
Supplier:  ACROBIOSYSTEMS
Cynomolgus Her2, His Tag (HE2-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession XP005584 091.2). 1 * 100 µG
Catalog # ACRBHE2-C52HB100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together