Orders LinkContactUsLinkComponent
Dosing spoons, white
Dosing spoons, white
Large package, 2,5 ml
Catalog # 300-0366
Supplier:   BURKLE
Dosing spoons, white
Catalog # 300-0366
Supplier:  BURKLE
Dosing spoons, white
Dosing spoons, white
Large package, 2,5 ml
Price
43,50 €
Log in for availability
1 * 100 ST
Product Details & Documents
Volumetric and dosing spoons can be used to measure small amounts of powder, granulates and liquids exactly and dose them precisely. For precise measuring, excess medium is stripped off the upper edge of the spoon. The content volume is engraved on the handle.
  • PS, white
  • Optionally packed individually and sterilised
  • Stripper edge
  • For single use
  • Cleanroom manufactured (cleanroom Class 7)
The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom or getting into the corners of a container, thus allowing for efficient emptying of small remaining amounts. The flat base makes it possible to set the volumetric spoon down without spilling the contents.
The disposable dosing spoon is suitable for use as a sample spoon for taking small sample quantities. Since this plastic volumetric spoon was developed for single use, there is no cleaning, and the possibility of cross-contamination and impurities is excluded.
These dosing and volumetric spoons are suitable for precise measurements in chemical and pharmaceutical applications, cosmetics, animal feed also the food and beverage industry.
Certifications: Complies with EU food requirements and FDA regulations.
Packaging: Available either in a large pack of 100 pieces or 100 pieces packed individually and radiation sterilised.
Specifications
  • Capacity:
    2,5 ml
  • Description:
    Large package