Orders LinkContactUsLinkComponent
Forceps, disposable
Forceps, disposable
Pointed, straight, +, Individually
Catalog # 300-0346
Supplier:   BURKLE
Forceps, disposable
Catalog # 300-0346
Supplier:  BURKLE
Forceps, disposable
Forceps, disposable
Pointed, straight, +, Individually
Price
93,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
The disposable forceps are suitable for precise lab work and are ideal for grasping, lifting and moving small objects, for example fine tissue, small items or solid samples.
  • PS, white
  • Can be individually packed and sterilised
  • For single use
  • Clean room manufactured (Class 7 cleanroom)
  • Overall length 130 mm
The fluted handles ensure optimum and secure handling as well as precise work. Light plastic forceps for single use allow for fatigue-free work. Cleanroom manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined.
These lab forceps are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses.
Pointed/angled shape: These disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends.
Pointed shape: These anatomical forceps have a straight tip, fluted on the inside making it possible to grip sensitive materials reliably.
Broad tip shape: These disposable forceps have ends with a broad tip. The broad tip is smooth and blunt preventing material from being damaged when gripped.
Certifications: Complies with EU food requirements and FDA regulations.
Packaging: Available either in a large pack with 100 pieces or 100 pieces packed individually and radiation sterilised.
Specifications
  • Packed:
    Individually
  • Sterile:
    +
  • Type:
    Pointed, straight
  • Seq:
    04