Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Extra fine Graefe forceps

Stainless steel, 100 mm, Smooth, Curved, 0,5×0,5 mm, 0,5 mm
  FSCI11160-10
Marketsource
Extra fine Graefe forceps
  FSCI11160-10
Marketsource
Extra fine Graefe forceps
Extra fine Graefe forceps

Stainless steel, 100 mm, Smooth, Curved, 0,5×0,5 mm, 0,5 mm

 
52,00 €
   
1 * 1 ST
 
 
  • Length:
    100 mm
  • Material:
    Stainless steel
  • Seq:
    03
  • Tip style:
    Smooth
  • Tip size:
    0,5×0,5 mm
  • Tip width:
    0,5 mm
  • Tip shape:
    Curved