Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Bag waste disposal, 72l , biohazard , HD-PE, with indicator field , 700 x 1100 x 0.050 mm 1 * 350 items
Catalog # RATI7003130
Supplier:
RATIOLAB
Expand image
Bag waste disposal, 72l , biohazard , HD-PE, with indicator field , 700 x 1100 x 0.050 mm 1 * 350 items
Catalog # RATI7003130
Supplier:
RATIOLAB
Frequently Bought Together
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Supplier: EPPENDORF
epT.I.P.S.® Bulk - Pipette Tips
Supplier: EPPENDORF
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR® Disposable Spatulas, PP
Supplier: VWR Collection
VWR®, Pipette Tips, Universal
Supplier: VWR Collection
VWR®, Pipette Tips, Standard
Supplier: VWR Collection
Combitips® advanced Eppendorf Quality Positive Displacement Pipette Tips
Supplier: EPPENDORF
VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
VWR®, Disposable Weighing Boats
Supplier: VWR Collection
Jerricans, narrow neck, HDPE, transparent or blue
Supplier: Kautex
VWR® Reagent Reservoirs
Supplier: VWR Collection