Tube,Microcentrifuge , Safety-Cap ,PP, 5.0 ml, natural 1 * 100 items
Catalog # RATI5615158
Supplier: RATIOLAB
Tube,Microcentrifuge , Safety-Cap ,PP, 5.0 ml, natural 1 * 100 items
Catalog # RATI5615158
Supplier: RATIOLAB
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
Labelling tapes, set
Supplier: PRECISION DYNAMICS TIMEM
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Cell culture salts, salt solutions and buffers
Supplier: Biowest
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection