Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sponge, VertiKlean®

Sponge, VertiKlean®

Light yellow, 130×130 mm, 15 mm, 1 sponge/bag, 120 bags/case
Catalog # 115-3203
Supplier:   CONTEC
Sponge, VertiKlean®
Catalog # 115-3203
Supplier:  CONTEC
Sponge, VertiKlean®
Sponge, VertiKlean®

Light yellow, 130×130 mm, 15 mm, 1 sponge/bag, 120 bags/case

Price
2 140,00 €
Log in for availability
1 * 120 ST
Product Details & Documents

The VertiKlean Sponge is a disposable laboratory sponge. It consists of Polynit wiping material that has been laminated to a clean polyester foam. The sponge is laser cut to completely seal the edges. It is designed for general cleaning and wiping but can also be used in laboratories.

  • High sorbency polyester foam with the abrasion resistance of Polynit fabric
  • Autoclavable
Specifications
  • Packed:
    1 sponge/bag, 120 bags/case
  • Colour:
    Light yellow
  • L×W:
    130×130 mm
  • Thickness:
    15 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.