Order Entry
Belgium
Orders LinkContactUsLinkComponent
epT.I.P.S.® Racks - Pipette Tips

epT.I.P.S.® Racks - Pipette Tips

20 - 300 µl, 55 mm, Orange, 480 tips (5 racks×96 tips)
Catalog # 613-6753
   Low Manufacturing Impact
Supplier:   EPPENDORF
epT.I.P.S.® Racks - Pipette Tips
Catalog # 613-6753
Supplier:  EPPENDORF
   Low Manufacturing Impact
epT.I.P.S.® Racks - Pipette Tips
epT.I.P.S.® Racks - Pipette Tips

20 - 300 µl, 55 mm, Orange, 480 tips (5 racks×96 tips)

Price
63,00 €
Log in for availability
1 * 480 ST
Product Details & Documents

Eppendorf epT.I.P.S.® Racks feature high-precision pipette tips in 'Eppendorf Quality', 'PCR clean', 'Biopur®', and 'Sterile' purity grades. All tips are supplied in disposible racks designed for optimal organisation and user convenience in demanding laboratory environments.

  • Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
  • Modern slim rack design with reclosable lid
  • Improved sustainability through 35% lower polypropylene consumption per rack
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability

The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally integrated pipetting system') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.

How to recognise an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.

Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0,5 to 20 µl, 50 to 1250 µl, 0,2 to 5 ml, and 0,5 to 10 ml.

 : Lot-specific certficates available for the purity grades PCR clean, Sterile, and Biopur® can be downloaded from the Eppendorf website.

Specifications
  • Colour code:
    Orange
  • Packed:
    480 tips (5 racks×96 tips)
  • Volume:
    20 - 300 µl
  • Length:
    55 mm
  • Seq:
    07
  • Cat. No.:
    613-6753
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact