ContactUsLinkComponent
Stainless steel basket for blood collection, basket with racks, A-295, rack for 64, 10 ml tubes 95 x 15 mm. B-240, rack for 24, 5 ml tubes 75 x 12 mm. A-261, rack for 28, 5 ml tubes 50 x 15 mm flat bottom. CP-24 1 * 1 items
Catalog # DELTH-401
Supplier:  DELTALAB
Stainless steel basket for blood collection, basket with racks, A-295, rack for 64, 10 ml tubes 95 x 15 mm. B-240, rack for 24, 5 ml tubes 75 x 12 mm. A-261, rack for 28, 5 ml tubes 50 x 15 mm flat bottom. CP-24 1 * 1 items
Catalog # DELTH-401
Supplier:  DELTALAB
Frequently Bought Together