ContactUsLinkComponent
Led light cubes CY5 - EX:640/30 EM:690/50 DM:660 1 * 1 items
Catalog # ECHLCUBE4005
Supplier:  DISCOVER ECHO
Led light cubes CY5 - EX:640/30 EM:690/50 DM:660 1 * 1 items
Catalog # ECHLCUBE4005
Supplier:  DISCOVER ECHO