Orders LinkContactUsLinkComponent
Objectives for Rebel Microscopes
Plan achromat phase, N.A. 0,25, 10,8 mm
Catalog # ECHLRMAG0113
Supplier:   DISCOVER ECHO
Objectives for Rebel Microscopes
Catalog # ECHLRMAG0113
Supplier:  DISCOVER ECHO
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan achromat phase, N.A. 0,25, 10,8 mm
Price
299,00 €
Log in for availability
1 * 1 ST
Specifications
  • Magnification:
    10×
  • Description:
    Plan achromat phase, N.A. 0,25
  • Seq:
    25
  • Working distance:
    10,8 mm