ContactUsLinkComponent
O. sensor element: GOEL 381, precise measuring at low O. (e.g. 35 vol. % O.) Measuring range oxygen concentration: 0 ... 100 % vol. O. (also for values <=0.2 % Vol. O.) Cable length: 1.2 m Working temperature: 0 ... 45 #‹C Servic 1 * 1 items
Catalog # GRSG610034
Supplier:  Greisinger
O. sensor element: GOEL 381, precise measuring at low O. (e.g. 35 vol. % O.) Measuring range oxygen concentration: 0 ... 100 % vol. O. (also for values <=0.2 % Vol. O.) Cable length: 1.2 m Working temperature: 0 ... 45 #‹C Servic 1 * 1 items
Catalog # GRSG610034
Supplier:  Greisinger
Frequently Bought Together