Order Entry
Belgium
Orders LinkContactUsLinkComponent
Generators, Polytron® PT-DA/BEC (BIOTRONA)

Generators, Polytron® PT-DA/BEC (BIOTRONA)

Polytron® 12 mm generator, PT-DA 12/BEC-E157 (BIOTRONA), PT 1200 E, PT 1300 D, PT 2500 E, 157 mm, 50 - 500 ml
Catalog # KINE11030286
Supplier:   KINEMATICA
Generators, Polytron® PT-DA/BEC (BIOTRONA)
Catalog # KINE11030286
Supplier:  KINEMATICA
Generators, Polytron® PT-DA/BEC (BIOTRONA)
Generators, Polytron® PT-DA/BEC (BIOTRONA)

Polytron® 12 mm generator, PT-DA 12/BEC-E157 (BIOTRONA), PT 1200 E, PT 1300 D, PT 2500 E, 157 mm, 50 - 500 ml

Price
2 240,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

BIOTRONA® design high turbulence mixing. High-turbulence mixing head with low shear forces and energy inputs. Time saving with improved results compared to stirrers and dissolvers of up to 90%. Dispersing generators are made from high-alloy stainless steel 316 L, electropolished, Ra≤1,6 μm as standard.

  • Fast dissolving and suspending of solid particles into liquids
  • Ideal for higher viscosities
  • Lower shear forces
Specifications
  • Volume:
    50 - 500 ml
  • Description:
    Polytron® 12 mm generator, PT-DA 12/BEC-E157 (BIOTRONA)
  • Length:
    157 mm
  • Seq:
    02
  • For:
    PT 1200 E, PT 1300 D, PT 2500 E
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.