ContactUsLinkComponent
Rifampicin 5 mg Disc. Each package should include 50 disc and each Disc should include rifampicin 5 mg 1 * 250 items
Catalog # HIMESD030-1PK
Supplier:  HIMEDIA
Rifampicin 5 mg Disc. Each package should include 50 disc and each Disc should include rifampicin 5 mg 1 * 250 items
Catalog # HIMESD030-1PK
Supplier:  HIMEDIA
Frequently Bought Together