Orders LinkContactUsLinkComponent
Multirack bulk tips, Axygen®
Multirack bulk tips, Axygen®
Extended length, 50 - 1250 µl
Catalog # AXYGMR-1000XT
Supplier:   Corning
Multirack bulk tips, Axygen®
Catalog # AXYGMR-1000XT
Supplier:  Corning
Multirack bulk tips, Axygen®
Multirack bulk tips, Axygen®
Extended length, 50 - 1250 µl
Price
391,00 €
Log in for availability
1 * 7.680 ST
Product Details & Documents
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an inter-changeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Nonpyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
Axygen® MultiRack bulk pipette tips are compatible with MultiRack racks and packaged in resealable polyethylene bags, non sterile.
Ordering information: Axygen® MultiRack pipette tips are also available in hinged rack, filter tips, reload, and tipstation configurations.
Specifications
  • Volume:
    50 - 1250 µl
  • Seq:
    03
  • Tip style:
    Extended length