ContactUsLinkComponent
HWS-PS cover, NW 100, Mh NS 29/32 Sh 1 x NS 29/32 g (3 o'clock) , 1 x GL18g (0 o'clock) 1 x NS29/32s (6 o'clock), 1 x NS29/32s (horn ) (9 o'clock) acc. to customer specification 1 * 1 items
Catalog # HWSA3310599/049
Supplier:  HWS LABORTECHNIK
HWS-PS cover, NW 100, Mh NS 29/32 Sh 1 x NS 29/32 g (3 o'clock) , 1 x GL18g (0 o'clock) 1 x NS29/32s (6 o'clock), 1 x NS29/32s (horn ) (9 o'clock) acc. to customer specification 1 * 1 items
Catalog # HWSA3310599/049
Supplier:  HWS LABORTECHNIK
Frequently Bought Together