Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Stirring rods

Stirring rods, glass, with fused ends, Glass, 9 mm, 300 mm
Catalog # HECH42850078
Stirring rods
Catalog # HECH42850078
Stirring rods
Stirring rods

Stirring rods, glass, with fused ends, Glass, 9 mm, 300 mm

Price
40,90 €
Log in for availability
1 * 25 ST
Product Details & Documents

AR-Glas®. With fused ends.

Specifications
  • Ø:
    9 mm
  • Description:
    Stirring rods, glass, with fused ends
  • Length:
    300 mm
  • Material:
    Glass
  • Seq:
    023
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.