Orders LinkContactUsLinkComponent
Accessories
Accessories
A 313 Module, suitable for laboratory flasks up to 10 L, round flasks up to 6 L, Erlenmeyer flasks up to 5 L, equipped with 5 injector positions, W×D×H: 200×320×20 mm
Catalog # MIEL11055970
Supplier:   MIELE
Accessories
Catalog # MIEL11055970
Supplier:  MIELE
Accessories
Accessories
A 313 Module, suitable for laboratory flasks up to 10 L, round flasks up to 6 L, Erlenmeyer flasks up to 5 L, equipped with 5 injector positions, W×D×H: 200×320×20 mm
Price
905,35 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    A 313 Module, suitable for laboratory flasks up to 10 L, round flasks up to 6 L, Erlenmeyer flasks up to 5 L, equipped with 5 injector positions, W×D×H: 200×320×20 mm
  • Seq:
    22