ContactUsLinkComponent
Microscope slides, 46x27 mm, thickness 2 mm, float glass, edges ground at 90, without frosted end, in boxes of 25 pcs., MARIENFELD SUPERIOR 1 * 1.000 items
Catalog # MARI1136491091
Supplier:  MARIENFELD
Microscope slides, 46x27 mm, thickness 2 mm, float glass, edges ground at 90, without frosted end, in boxes of 25 pcs., MARIENFELD SUPERIOR 1 * 1.000 items
Catalog # MARI1136491091
Supplier:  MARIENFELD
Frequently Bought Together