ContactUsLinkComponent
Microscope slides, 76x26 mm, thickness 1 mm, pure white glass, edges ground at 90, without frosted end, paper-interleaved, in boxes of 50 pcs., 2500 pcs./carton, CE marked 1 * 2.500 items
Catalog # MARI1010419091
Supplier:  MARIENFELD
Microscope slides, 76x26 mm, thickness 1 mm, pure white glass, edges ground at 90, without frosted end, paper-interleaved, in boxes of 50 pcs., 2500 pcs./carton, CE marked 1 * 2.500 items
Catalog # MARI1010419091
Supplier:  MARIENFELD
Frequently Bought Together