ContactUsLinkComponent
CE certi?cation: this clog is a personal protective equipment in accordance to the requirements of working shoes Light: average weight 700 gr. per pair Aerated clog: lateral holes provide aeration but avoid liquids to drop in falling From height Anat 1 * 1 PAIR
Catalog # BSNCGL040/38
Supplier:  Basan
CE certi?cation: this clog is a personal protective equipment in accordance to the requirements of working shoes Light: average weight 700 gr. per pair Aerated clog: lateral holes provide aeration but avoid liquids to drop in falling From height Anat 1 * 1 PAIR
Catalog # BSNCGL040/38
Supplier:  Basan
Frequently Bought Together