Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, Wire baskets

VWR®, Wire baskets

200×300×200 mm
Catalog # 215-4206
Supplier:   VWR Collection
VWR®, Wire baskets
Catalog # 215-4206
Supplier:  VWR Collection
VWR®, Wire baskets
VWR®, Wire baskets

200×300×200 mm

Price
274,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These wire baskets are made from rust-proof, electrolytically polished stainless steel.

  • Very good chemical and temperature resistance
  • Spot-welded mesh 8×8×1 mm
  • Reinforced edges

Can be used for transport, storage or sterilisation of laboratory utensils.

Specifications
  • W×D×H:
    200×300×200 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.