ContactUsLinkComponent
Chromogenic media on double plate for urinary bacterial count and identification and for detecting Extended Spectrum Beta-Lactamase producing bacteria. 1 * 20 items
Catalog # LIOF18011
Supplier:  LIOFILCHEM
Chromogenic media on double plate for urinary bacterial count and identification and for detecting Extended Spectrum Beta-Lactamase producing bacteria. 1 * 20 items
Catalog # LIOF18011
Supplier:  LIOFILCHEM
Frequently Bought Together