Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

SafetyCaps

SafetyCap II, V2.0, GL45, 2× PFA-fitting 1,6/2,3/3,2 mm OD
Catalog # SCTV307909
Supplier:   S.C.A.T.
SafetyCaps
Catalog # SCTV307909
Supplier:  S.C.A.T.
SafetyCaps
SafetyCaps

SafetyCap II, V2.0, GL45, 2× PFA-fitting 1,6/2,3/3,2 mm OD

Price
112,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The SCAT SafetyCaps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapours. Complete with integrated air valve for pressure equalisation.

  • High quality materials ensure maximum safety for lab and staff
  • 360° rotatable and up to 200 °C autoclavable screw cap for maximum flexibility
  • Stable construction for highest safety and easy handling
  • Chemically resistant against aggressive organic solvents
  • Attractive design and clear arrangement of the connections
  • Improved design for easy connection of capillaries in various colours
  • Free air vaive with change indicator included with every SafetyCap

SCAT SafetyCaps are available in the following versions:

SafetyCaps, basic: Standard SafetyCap with different connections without additional functions.
SafetyCaps, preparative: For use in the preparative HPLC environment with larger tubing connections and air valves with a higher flow rate.
SafetyCaps, with shut-off: Prevent air pockets in the tube when the supply bottle is being changed and guarantees that the analysis continues without a problem.
SafetyCaps, combined: Provides a stable baseline and reliable reproducibility for RI detection for a fast and simple solvent degassing with helium.
SafetyCaps, angled: With lateral connections for capillary and air valve in case of limited space above the bottle.
SafetyCaps, for Merck® bottles: Connect containers with thread sizes S 40 and GL 40 directly to the HPLC system, without decanting or thread adapters.
SafetyCaps, for NS 29/32 ground neck bottles: The locknut allows the stopper to be easily loosened and removed for bottles with NS 29/32 mm standard ground joints.

 : All SafetyCaps are delivered with an air valve and corresponding fittings. PFA fittings, PTFE body and PPS screw cap are conforming to the requirements of flammability classification V-0 (UL-94). Blind plugs are provided to tighten connections which are temporarily inactive or not in use.

Specifications
  • Thread:
    GL45
  • Description:
    SafetyCap II, V2.0
  • Seq:
    02
  • Connector:
    2× PFA-fitting 1,6/2,3/3,2 mm OD