ContactUsLinkComponent
HF5-EX TRANSMITTER duct version with display 2-wires 4...20mA voltage supply: 10...28 VDC analogue outputs: OUT1: 4...20mA = 0...100%RH OUT2: 4...20mA = -30...+70C 1 * 1 items
Catalog # ROTRHF520EXDB4XD1X
HF5-EX TRANSMITTER duct version with display 2-wires 4...20mA voltage supply: 10...28 VDC analogue outputs: OUT1: 4...20mA = 0...100%RH OUT2: 4...20mA = -30...+70C 1 * 1 items
Catalog # ROTRHF520EXDB4XD1X
Frequently Bought Together