Bis(NHS)PEG5
100 mg
Catalog # TCIAB3728-100MG
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: 4,7,10,13,16-Pentaoxanonadecanedioic Acid Di(N-succinimidyl) Ester, Bis(2,5-dioxopyrrolidin-1-yl) 4,7,10,13,16-pentaoxanonadecane-1,19-dioate, Di(N-succinimidyl) 4,7,10,13,16-Pentaoxanonadecanedioate
- CAS Number:756526-03-1
- Formula:C₂₂H₃₂N₂O₁₃
- MDL Number:MFCD11041118
- Molecular Weight:532.50 g/mol
- Storage Temperature:Freezer
Documents
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid to cloudy liquid
- Elemental analysis (Nitrogen):4.70 to 5.80 %
Specifications
- Pk:100 mg



