14 components 100µg/kg each of Ge Nb Pd Pt Rh Sb Sn Ta Ti W Zr Ir Os Ru in HNO3 5% HCl 0.5% HF 0.5% . 1 * 100 mL
Catalog # ACSDRM030144L1
Supplier: CPACHEM
14 components 100µg/kg each of Ge Nb Pd Pt Rh Sb Sn Ta Ti W Zr Ir Os Ru in HNO3 5% HCl 0.5% HF 0.5% . 1 * 100 mL
Catalog # ACSDRM030144L1
Supplier: CPACHEM
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals