Orders LinkContactUsLinkComponent
Fluorite Objectives for Revolve Microscope
Fluorite Objectives for Revolve Microscope
U-Plan fluorite objective, 0,2 mm
Catalog # ECHLJMAG0609
Supplier:   DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Catalog # ECHLJMAG0609
Supplier:  DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
U-Plan fluorite objective, 0,2 mm
Price
3 990,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
    60×
  • Description:
    U-Plan fluorite objective
  • Seq:
    06
  • Working distance:
    0,2 mm