VWR®, Sampling Scoops
25 ml, 141 mm, White
Supplier: VWR Collection
Jump to:
Product Details & Documents
PS, white. These sterile scoops are ideal for sampling powder, granules and pastes.
- Individually packed
- Sterilised by gamma radiation
Thanks to the special handle construction, and the slightly angled edge, the scoop stands horizontally and can be laid down without the material trickling out.
: Production, assembly and packaging according to cleanroom Class 7 (10000). According to EU food and FDA guidelines.
: *The product(s) below are outside the scope of the EU Directive 2019/904.
Documents
Specifications
- Capacity:25 ml
- Colour:White
- Length:141 mm
- Seq:01
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



