IONIC BUFFER TISAB IV 1 * 5 L
PRLS89330.360
Frequently Bought Together
Beakers, triangular, Tricorn™
: Simport Scientific
NEOMYCIN ELISA KIT 1 * 1 KIT
: ENZO LIFE SCIENCES
SOLUTION ACIDE NITRIQUE 0.5N 1M NAH2PO4 1 * 1 L
: PROLABO CMP
[EN]IGG4 ISOTYPE CTRL S228PF234AL235A LA 1 * 1 mg
: MARKET SOURCE PART PROCESS
Malachite green oxalate, pure, certified
: Thermo Fisher Scientific
CALIBRATOR 26 1 * 1 Vial
: MESO SCALE DISCOVERY
1 COMPONENT: HG 0.5MG/L IN HCL 5% 1 * 250 mL
: CUSTOM MADE CHEMICALS LAB
POTASSIUM CYANURE EN SOLUTION 1 * 100 mL
: PROLABO CMP