neoLab Cannulas Luer-Lock-Anschluss, 0,80 x 120 mm 1 * 3 ST
Catalog # NEOL2-3107
Fournisseur: NEOLAB
neoLab Cannulas Luer-Lock-Anschluss, 0,80 x 120 mm 1 * 3 ST
Catalog # NEOL2-3107
Fournisseur: NEOLAB
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Fournisseur: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Fournisseur: VWR Chemicals
Acetone ≥99%, TECHNICAL
Fournisseur: VWR Chemicals
Sealing film for manual application, Parafilm® M
Fournisseur: HEATHROW
VWR® Facial Tissues
Fournisseur: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Fournisseur: VWR Chemicals
VWR®, Nitrile Gloves
Fournisseur: VWR Collection
pH indicator strips, MQuant®, Supelco®
Fournisseur: Merck
VWR® NITRILE LIGHT, Nitrile Gloves
Fournisseur: VWR Collection
VWR®, Transfer Pipettes, High Performance
Fournisseur: VWR Collection
VWR® Serological Pipette, Standard Line
Fournisseur: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Fournisseur: KIMBERLY CLARK