CellStor Pot™ specimen container
60 ml, 10% neutral buffered formalin
Catalog # CLLPBAF-6000-08A
Supplier: CELLPATH
Jump to:
Product Details & Documents
Prefilled specimen containers.
- Leak-proof lid for safe transportation and specimen storage
- With traceable badge number, safety data sheet in the packaging
Specifications
- Capacity:60 ml
- Description:10% neutral buffered formalin
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



