Objectives for Rebel Microscopes
Plan apochromat, N.A. 0,95, 0,15 mm
Catalog # ECHLRMAG9402
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
Specifications
- Magnification:40×
- Description:Plan apochromat, N.A. 0,95
- Seq:03
- Working distance:0,15 mm