Orders LinkContactUsLinkComponent
FLED fluorescence intermediate attachments
FLED fluorescence intermediate attachments
FLED Fluorescence intermediate attachment (TRITC)
- Fluorescence intermediate part
- LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
- Light blocking slider
Catalog # MOTI1101011400201
Supplier:   MOTIC
FLED fluorescence intermediate attachments
Catalog # MOTI1101011400201
Supplier:  MOTIC
FLED fluorescence intermediate attachments
FLED fluorescence intermediate attachments
FLED Fluorescence intermediate attachment (TRITC)
- Fluorescence intermediate part
- LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
- Light blocking slider
Price
2 250,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Compatible with Panthera U, KU and CC microscopes.
Specifications
  • Description:
    FLED Fluorescence intermediate attachment (TRITC)
    - Fluorescence intermediate part
    - LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
    - Light blocking slider