Orders LinkContactUsLinkComponent
FLED fluorescence intermediate attachments
FLED fluorescence intermediate attachments
FLED Fluorescence intermediate attachment (Auramine O)
- Fluorescence intermediate part
- LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
- Light blocking slider
Note: Recommended to use with Plan objectives UC PL NCG
Catalog # MOTI1101011400181
Supplier:   MOTIC
FLED fluorescence intermediate attachments
Catalog # MOTI1101011400181
Supplier:  MOTIC
FLED fluorescence intermediate attachments
FLED fluorescence intermediate attachments
FLED Fluorescence intermediate attachment (Auramine O)
- Fluorescence intermediate part
- LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
- Light blocking slider
Note: Recommended to use with Plan objectives UC PL NCG
Price
2 580,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Compatible with Panthera U, KU and CC microscopes.
Specifications
  • Description:
    FLED Fluorescence intermediate attachment (Auramine O)
    - Fluorescence intermediate part
    - LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
    - Light blocking slider
    Note: Recommended to use with Plan objectives UC PL NCG