HyClone™ FetalClone™ I serum, U.S. origin
FetalClone® I, US origin, irradiated 25 - 40 kGy, Gamma irradiated, Frozen
Catalog # SH30080.03IR25
Supplier: HyClone products (Cytiva)
HyClone™ FetalClone™ I serum, U.S. origin
Catalog # SH30080.03IR25
Supplier: HyClone products (Cytiva)
Jump to:
Product Details & Documents
HyClone™ FetalClone® I is a foetal bovine serum alternative optimised for the growth of hybridomas with comparable IgG levels to those found in FBS.
- IgG levels comparable with those found in FBS
- Demonstrated cell growth performance equivalency to FBS
- High lot-to-lot consistency
- Low endotoxin and hemoglobin levels
Sterile filtered using three sequential 100 nm (0.1 µm) pore-size rated filters. Each lot undergoes extensive quality performance and analytical testing.
Documents
Certificates
Search by Lot NumberSpecifications
- Serum format:Frozen
- Sterility:Gamma irradiated
- Description:FetalClone® I, US origin, irradiated 25 - 40 kGy
- Seq:06
- Cat. No.:SH30080.03IR25
- Storage temperature:Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



