Orders LinkContactUsLinkComponent
VWR®, Cell Holders
VWR®, Cell Holders
Cell holder, water-jacketed 4-position 10 mm cell holder, used for P4, PV4, M4, P6, M6
Catalog # 634-1134
Supplier:   VWR Collection
VWR®, Cell Holders
Catalog # 634-1134
Supplier:  VWR Collection
VWR®, Cell Holders
VWR®, Cell Holders
Cell holder, water-jacketed 4-position 10 mm cell holder, used for P4, PV4, M4, P6, M6
Price
566,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Cell holder, water-jacketed 4-position 10 mm cell holder, used for P4, PV4, M4, P6, M6
  • Seq:
    12