Orders LinkContactUsLinkComponent
Conductivity standards, Supelco®
Conductivity standards, Supelco®
Conductivity standard, 25 µmhos/cm, 500 mL
Catalog # COND25-500ML
Supplier:   Merck
Conductivity standards, Supelco®
Catalog # COND25-500ML
Supplier:  Merck
CAS Number:  
Conductivity standards, Supelco®
Conductivity standard, 25 µmhos/cm, 500 mL
CAS Number:  
Price
101,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
100 µmhos/cm = 100 µS/cm
Certifications: These certified reference materials (CRMs) ares produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Documents
Specifications
Specifications
  • Pk:
    500 mL
  • Description:
    Conductivity standard
  • Conductivity:
    25 µmhos/cm