Big-Star-Desiccator Low, Aluminium frame with panels made of acrylic glass, including one shelf made of stainless steel, tray and hy grometer, usable with up to 5 shelves, stackable. 1 * 1 items
Catalog # BOHLV1897-07
Supplier: Bohlender
Frequently Bought Together